Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC17A3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001098486.1 | NP_001091956.1 | 9 | 1221 | ||
NM_006632.3 | NP_006623.2 | 8 | 987 | |||
GenBank mRNA | AK296070.1 | - | 5 | 1364 | ||
AK298271.1 | - | 9 | 1221 | |||
AK298297.1 | - | 5 | 1309 | |||
BC017952.1 | - | 8 | 956 | 4557795 | ||
CB163247.1 | - | 3 | 359 | |||
U90545.1 | - | 8 | 1196 |