Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bcar3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013867.2 | NP_038895.1 | 5 | 996 | ||
GenBank mRNA | AF179566.1 | - | 5 | 944 | ||
AK143894.1 | - | 5 | 784 | |||
AK158241.1 | - | 5 | 998 | |||
AK167578.1 | - | 5 | 815 | |||
BC023930.1 | - | 5 | 985 | 5324365 |