Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rnf11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013876.3 | NP_038904.1 | 3 | 822 | ||
GenBank mRNA | AB024427.1 | - | 4 | 687 | ||
AK014254.1 | - | 3 | 975 | |||
BC010299.1 | - | 3 | 822 | 3155804 | ||
BC028255.1 | - | 3 | 1003 | 4972336 |