Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SDCCAG8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006642.3 | NP_006633.1 | 10 | 1362 | ||
XM_005273013.4 | XP_005273070.1 | 9 | 1194 | |||
XM_005273018.1 | XP_005273075.1 | 9 | 1201 | |||
XM_005273021.4 | XP_005273078.1 | 10 | 1134 | |||
XM_005273022.3 | XP_005273079.1 | 7 | 983 | |||
XM_006711727.3 | XP_006711790.1 | 10 | 1248 | |||
XM_006711728.3 | XP_006711791.1 | 9 | 1119 | |||
XM_011544021.2 | XP_011542323.1 | 11 | 1344 | |||
XM_011544022.2 | XP_011542324.1 | 11 | 1419 | |||
XM_011544023.2 | XP_011542325.1 | 11 | 1344 | |||
XM_011544024.2 | XP_011542326.1 | 11 | 1344 | |||
XM_011544026.2 | XP_011542328.1 | 11 | 1344 | |||
XM_011544028.2 | XP_011542330.1 | 9 | 1119 | |||
XM_011544030.2 | XP_011542332.1 | 3 | 1895 | |||
XM_017000103.1 | XP_016855592.1 | 11 | 1344 | |||
XM_017000104.1 | XP_016855593.1 | 9 | 1194 | |||
XM_017000105.1 | XP_016855594.1 | 10 | 1349 | |||
XM_017000106.1 | XP_016855595.1 | 10 | 1330 | |||
XM_017000107.1 | XP_016855596.1 | 11 | 1246 | |||
XM_017000108.1 | XP_016855597.1 | 13 | 1793 | |||
GenBank mRNA | AF039690.1 | - | 4 | 533 | ||
AF250731.1 | - | 10 | 1193 | |||
AK297105.1 | - | 6 | 781 | |||
AK299249.1 | - | 7 | 954 |