Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Timm8a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013898.2 | NP_038926.1 | 2 | 541 | ||
GenBank mRNA | AB031055.1 | - | 2 | 541 | ||
AF150081.1 | - | 2 | 532 | |||
AK011402.1 | - | 2 | 576 | |||
AK088903.1 | - | 2 | 534 | |||
AK090079.1 | - | 2 | 580 | |||
AK165573.1 | - | 2 | 536 | |||
BC004796.1 | - | 2 | 529 | 3588797 | ||
BC094631.1 | - | 2 | 550 | 3419065 |