Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Usp21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013919.4 | NP_038947.2 | 5 | 1058 | ||
GenBank mRNA | AF177759.1 | - | 4 | 857 | ||
AK011148.1 | - | 5 | 1068 | |||
AK013252.1 | - | 5 | 1053 | |||
AK049649.1 | - | 3 | 1056 | |||
BC021903.1 | - | 5 | 1048 | 3985792 | ||
KF646669.1 | - | 4 | 576 |