Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Shroom3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001077595.2 | NP_001071063.1 | 4 | 1314 | ||
NM_001077596.2 | NP_001071064.1 | 4 | 1362 | |||
NM_015756.2 | NP_056571.2 | 5 | 1686 | |||
GenBank mRNA | AF199421.1 | - | 5 | 1686 | ||
AF199422.1 | - | 4 | 1254 | |||
AK129371.1 | - | 1 | 692 | |||
AK147493.1 | - | 4 | 1297 | |||
AK164918.1 | - | 4 | 1273 | |||
BC145113.1 | - | 5 | 1496 | 9053661 |