Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ebi3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015766.2 | NP_056581.1 | 3 | 540 | ||
GenBank mRNA | AF013114.1 | - | 3 | 523 | ||
AK155023.1 | - | 3 | 503 | |||
AK155903.1 | - | 3 | 395 | |||
BC008209.1 | - | 3 | 488 | 3154957 | ||
CX240802.1 | - | 3 | 487 |