Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pdcd7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016688.2 | NP_057897.2 | 5 | 2259 | ||
GenBank mRNA | AF083929.1 | - | 5 | 2252 | ||
AK004096.1 | - | 3 | 1052 | |||
AK136141.1 | - | 4 | 880 | |||
AK155050.1 | - | 5 | 2224 | |||
BC022772.1 | - | 4 | 1296 |