Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Map3k6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016693.5 | NP_057902.5 | 5 | 995 | ||
GenBank mRNA | AB021861.4 | - | 5 | 993 | ||
BC120565.1 | - | 5 | 922 | 40129488 | ||
BC125577.1 | - | 5 | 922 | 40130092 |