Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Preb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001294302.1 | NP_001281231.1 | 5 | 942 | ||
NM_016703.3 | NP_057912.2 | 5 | 942 | |||
GenBank mRNA | AF150808.1 | - | 5 | 845 | ||
AF193017.1 | - | 5 | 854 | |||
AK013330.1 | - | 5 | 896 | |||
AK029838.1 | - | 5 | 878 | |||
AK036866.1 | - | 5 | 880 | |||
AK151288.1 | - | 5 | 924 | |||
AK188795.1 | - | 3 | 256 | |||
AK200030.1 | - | 3 | 257 | |||
AK213634.1 | - | 3 | 257 | |||
BC017527.1 | - | 5 | 845 | 4921119 |