Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Atp2a3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163336.1 | NP_001156808.1 | 13 | 1938 | ||
NM_001163337.1 | NP_001156809.1 | 13 | 1938 | |||
NM_016745.3 | NP_058025.2 | 13 | 1938 | |||
GenBank mRNA | AK089487.1 | - | 13 | 1938 | ||
AK155904.1 | - | 13 | 1939 | |||
BC017639.1 | - | 13 | 1852 | 4924519 | ||
BC026147.1 | - | 13 | 1884 | 4191458 | ||
U49393.1 | - | 13 | 1799 | |||
U49394.1 | - | 13 | 1799 |