Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Folh1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001159706.1 | NP_001153178.1 | 6 | 882 | ||
NM_016770.3 | NP_058050.3 | 6 | 882 | |||
GenBank mRNA | AF026380.1 | - | 6 | 864 | ||
AK002920.1 | - | 6 | 849 | |||
AK090289.1 | - | 883 | ||||
BC119604.1 | - | 6 | 826 | 40060385 | ||
BC119605.1 | - | 6 | 826 | 40060386 |