Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: COPS8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006710.4 | NP_006701.1 | 8 | 1683 | ||
NM_198189.2 | NP_937832.1 | 9 | 1748 | |||
GenBank mRNA | AK289891.1 | - | 9 | 1282 | ||
BC003090.1 | - | 8 | 1207 | 3503287 | ||
BC036499.1 | - | 8 | 1817 | 5264527 | ||
BQ716842.1 | - | 1 | 391 |