Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Uevld | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001040695.1 | NP_001035785.1 | 4 | 334 | ||
GenBank mRNA | AK086750.1 | - | 2 | 1069 | ||
AK154536.1 | - | 4 | 285 | |||
AK155685.1 | - | 4 | 281 | |||
BC054796.1 | - | 4 | 340 | 6843964 | ||
CB524764.1 | - | 4 | 340 | |||
CN538811.1 | - | 3 | 204 | |||
U97571.1 | - | 4 | 288 |