Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Krt85 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016879.2 | NP_058575.2 | 9 | 1931 | ||
GenBank mRNA | AF021836.1 | - | 9 | 1915 | ||
AK009099.1 | - | 9 | 1930 | |||
AK028825.1 | - | 9 | 1931 |
Gene Symbol: Krt83 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AA755006.1 | - | 1 | 348 |