Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Timm23 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016897.3 | NP_058593.2 | 7 | 840 | ||
GenBank mRNA | AB021122.1 | - | 7 | 820 | ||
AK013976.1 | - | 7 | 786 | |||
AK146516.1 | - | 7 | 806 | |||
BC037022.1 | - | 8 | 817 | 5366968 | ||
BC062963.1 | - | 7 | 776 | 5692352 | ||
BC069903.1 | - | 7 | 810 | 5290493 | ||
BC096669.1 | - | 7 | 809 | 5056242 | ||
BC132019.1 | - | 7 | 789 | 40130296 | ||
BC132319.1 | - | 7 | 789 | 40130596 |