Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Timm23 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016897.3 | NP_058593.2 | 7 | 1150 | ||
GenBank mRNA | AB021122.1 | - | 7 | 1130 | ||
BC037022.1 | - | 8 | 1127 | 5366968 | ||
BC069903.1 | - | 7 | 1120 | 5290493 | ||
BC096669.1 | - | 7 | 1119 | 5056242 |