Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: B4galt2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001253381.1 | NP_001240310.1 | 8 | 1617 | ||
NM_017377.5 | NP_059073.1 | 8 | 1582 | |||
GenBank mRNA | AF142670.1 | - | 7 | 1226 | ||
AK145250.1 | - | 9 | 1639 | |||
AK165829.1 | - | 8 | 1576 | |||
BC138567.1 | - | 7 | 1218 | 8861587 | ||
BC138569.1 | - | 7 | 1218 | 8861589 | ||
BC145335.1 | - | 7 | 1253 | 9053891 | ||
BX526030.1 | - | 1 | 69 |