Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Copg1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017477.2 | NP_059505.1 | 14 | 1501 | ||
GenBank mRNA | AF187079.1 | - | 14 | 1500 | ||
AK049417.1 | - | 14 | 1452 | |||
AK077461.1 | - | 14 | 1448 | |||
AK147895.1 | - | 14 | 1514 | |||
AK151284.1 | - | 14 | 1506 | |||
AK151816.1 | - | 14 | 1506 | |||
AK153142.1 | - | 14 | 1506 | |||
AK164280.1 | - | 14 | 1471 | |||
AK169256.1 | - | 14 | 1546 | |||
AK171950.1 | - | 14 | 1480 | |||
AK172519.1 | - | 14 | 1517 | |||
AK193696.1 | - | 1 | 46 | |||
AK194411.1 | - | 1 | 46 | |||
AK195632.1 | - | 1 | 47 | |||
AK197699.1 | - | 1 | 46 | |||
AK201780.1 | - | 1 | 46 | |||
AK204845.1 | - | 1 | 47 | |||
BC008553.1 | - | 6 | 690 | |||
BC024686.1 | - | 14 | 1472 | 3992144 | ||
BC024896.1 | - | 14 | 1469 | 4189311 | ||
BC060744.1 | - | 3 | 399 |