Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Abhd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018811.6 | NP_061281.3 | 10 | 1240 | ||
GenBank mRNA | AF201730.1 | - | 10 | 1229 | ||
AF546701.1 | - | 10 | 1241 | |||
AK008690.1 | - | 10 | 1250 | |||
AK016296.1 | - | 8 | 901 | |||
AK139526.1 | - | 10 | 1249 | |||
AK153642.1 | - | 10 | 1287 | |||
BC027098.1 | - | 10 | 1210 | 5356667 | ||
BC089477.1 | - | 10 | 1220 | 6741521 |