Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mpc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018819.4 | NP_061289.1 | 5 | 803 | ||
GenBank mRNA | AF181116.1 | - | 5 | 759 | ||
AK002241.1 | - | 5 | 804 | |||
AK002266.1 | - | 5 | 782 | |||
AK004013.1 | - | 5 | 759 | |||
AK005348.1 | - | 5 | 795 | |||
AK013391.1 | - | 5 | 804 | |||
AK014421.1 | - | 5 | 761 | |||
AK135943.1 | - | 5 | 774 | |||
AK170514.1 | - | 5 | 820 | |||
BC103767.1 | - | 5 | 806 | 30840199 |