Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Asah2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018830.1 | NP_061300.1 | 13 | 1563 | ||
GenBank mRNA | AB037111.1 | - | 14 | 1848 | ||
AB037181.1 | - | 13 | 1563 | |||
AK034493.1 | - | 13 | 1795 | |||
AK046540.1 | - | 13 | 1757 | |||
AK080951.1 | - | 13 | 1740 | |||
BC022604.1 | - | 4 | 345 | |||
BC137898.1 | - | 13 | 1546 | 8860917 | ||
BC145016.1 | - | 13 | 1549 | 9053564 |