Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Camkk1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018883.2 | NP_061371.2 | 16 | 3198 | ||
GenBank mRNA | AF117384.1 | - | 16 | 3146 | ||
AK144483.1 | - | 15 | 3468 | |||
BC017529.1 | - | 16 | 3163 | 4924656 |