Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mtmr7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001040699.1 | NP_001035789.1 | 2 | 181 | ||
GenBank mRNA | AK028160.1 | - | 2 | 146 | ||
AK081973.1 | - | 2 | 167 | |||
AK148768.1 | - | 2 | 182 | |||
BC032254.1 | - | 2 | 183 | 5368816 | ||
BY741806.1 | - | 2 | 182 |