Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ndufa1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019443.2 | NP_062316.1 | 3 | 442 | ||
GenBank mRNA | AK007840.1 | - | 3 | 349 | ||
AK131584.1 | - | 641 | ||||
BC024364.1 | - | 3 | 351 | 5031206 | ||
CF383819.1 | - | 2 | 226 | |||
Y07708.1 | - | 3 | 461 |