Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Btg4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019493.3 | NP_062366.2 | 4 | 585 | ||
GenBank mRNA | AB050983.2 | - | 4 | 585 | ||
AJ005120.1 | - | 4 | 458 | |||
AK136041.1 | - | 4 | 572 | |||
AK136124.1 | - | 4 | 573 | |||
AK136149.1 | - | 4 | 575 | |||
AK136185.1 | - | 4 | 577 | |||
AK161379.1 | - | 6 | 664 | |||
AK162122.1 | - | 4 | 572 | |||
AK163407.1 | - | 4 | 571 | |||
BC066811.1 | - | 4 | 559 | 30475849 |