Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ramp3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019511.3 | NP_062384.1 | 3 | 738 | ||
GenBank mRNA | AF146524.1 | - | 3 | 739 | ||
AF209907.1 | - | 3 | 698 | |||
AJ250491.1 | - | 3 | 739 | |||
AK005513.1 | - | 4 | 854 | |||
BC024765.1 | - | 3 | 734 | 5364778 |