Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rbm38 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019547.2 | NP_062420.2 | 4 | 1677 | ||
GenBank mRNA | AK052886.1 | - | 2 | 1462 | ||
AK146026.1 | - | 4 | 1651 | |||
AK166609.1 | - | 5 | 794 | |||
AK172625.1 | - | 4 | 1648 | |||
BC006687.1 | - | 3 | 2186 | |||
BC085307.1 | - | 4 | 1621 | 6396103 | ||
DV644612.1 | - | 1 | 156 |