Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tspan5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019571.5 | NP_062517.1 | 7 | 1070 | ||
GenBank mRNA | AF121344.1 | - | 7 | 1085 | ||
AK013350.1 | - | 7 | 1035 | |||
AK015705.1 | - | 7 | 792 | |||
AK033543.1 | - | 7 | 769 | |||
BC046784.1 | - | 7 | 909 | 6416107 | ||
BC058695.1 | - | 7 | 1041 | 6813872 |