Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Patz1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001253690.1 | NP_001240619.1 | 1 | 252 | ||
NM_001253691.1 | NP_001240620.1 | 1 | 252 | |||
NM_019574.3 | NP_062520.1 | 1 | 252 | |||
GenBank mRNA | AB029397.1 | - | 1 | 252 | ||
AK166189.1 | - | 1 | 373 | |||
BC043035.1 | - | 1 | 270 | 5697155 | ||
BC072649.1 | - | 2 | 1314 |