Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Clptm1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019649.2 | NP_062623.2 | 4 | 351 | ||
GenBank mRNA | AK148080.1 | - | 4 | 333 | ||
AK152410.1 | - | 4 | 341 | |||
AK152809.1 | - | 4 | 341 | |||
AK165742.1 | - | 4 | 351 | |||
AK171901.1 | - | 4 | 342 | |||
BC022172.1 | - | 4 | 342 | 4193279 | ||
BY288640.1 | - | 4 | 356 | |||
D67067.1 | - | 4 | 338 |