Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Socs5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019654.2 | NP_062628.2 | 3 | 1516 | ||
GenBank mRNA | AF033187.1 | - | 1 | 1074 | ||
AK172997.1 | - | 1 | 60 | |||
BC053015.1 | - | 3 | 1516 | 5702321 |