Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pias1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019663.3 | NP_062637.2 | 11, 12 | 1481 | ||
GenBank mRNA | AF077950.1 | - | 11, 12 | 1473 | ||
AK013753.1 | - | 7, 8 | 815 | |||
AK075708.1 | - | 11, 12 | 1482 | |||
AK153871.1 | - | 11, 12 | 1486 | |||
BC051417.1 | - | 11, 12 | 1478 | 6515027 |