Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tubd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199045.1 | NP_001185974.1 | 4 | 739 | ||
NM_001199046.1 | NP_001185975.1 | 4 | 633 | |||
NM_001199047.1 | NP_001185976.1 | 4 | 739 | |||
NM_019756.3 | NP_062730.1 | 4 | 633 | |||
GenBank mRNA | AB024329.1 | - | 3 | 407 | ||
AB031043.1 | - | 3 | 345 | |||
AF081568.1 | - | 4 | 473 | |||
AK016086.1 | - | 4 | 433 | |||
AK030214.1 | - | 4 | 739 | |||
AK169454.1 | - | 4 | 634 | |||
AK170304.1 | - | 4 | 598 | |||
AK171546.1 | - | 4 | 600 | |||
BC026465.1 | - | 4 | 616 | 4484099 | ||
BC132325.1 | - | 4 | 466 | 40130602 |