Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Akap8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AB028920.1 | - | 3, 4 | 126 | ||
AK004801.1 | - | 3, 4 | 132 | |||
AK089092.1 | - | 3, 4 | 139 | |||
BC042602.1 | - | 3, 4 | 122 | |||
BC060170.1 | - | 3, 4 | 117 | |||
BC060209.1 | - | 3, 4 | 223 | |||
BC078448.1 | - | 3, 4 | 110 | |||
BC125521.1 | - | 3, 4 | 122 | 40130036 | ||
BC137746.1 | - | 3, 4 | 122 | 8860765 |