Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cyp3a25 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019792.2 | NP_062766.2 | 11 | 1260 | ||
GenBank mRNA | AF204959.1 | - | 11 | 1259 | ||
AK149541.1 | - | 10 | 1173 | |||
BC028855.1 | - | 11 | 1251 | 5124151 | ||
Y11995.1 | - | 11 | 1262 |