Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tspan3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019793.3 | NP_062767.3 | 6 | 828 | ||
GenBank mRNA | AB009021.1 | - | 6 | 610 | ||
AB072450.1 | - | 6 | 638 | |||
AF133427.1 | - | 6 | 622 | |||
AF242591.1 | - | 6 | 821 | |||
AK006798.1 | - | 6 | 704 | |||
AK053325.1 | - | 6 | 679 | |||
AK089494.1 | - | 660 | ||||
AK141951.1 | - | 6 | 686 | |||
AK146130.1 | - | 6 | 816 | |||
AK152175.1 | - | 6 | 713 | |||
AK154919.1 | - | 660 | ||||
AK159324.1 | - | 6 | 799 | |||
AK159418.1 | - | 6 | 772 | |||
AK159621.1 | - | 6 | 783 | |||
AK159914.1 | - | 6 | 761 | |||
AK168231.1 | - | 6 | 816 | |||
AK169068.1 | - | 6 | 799 | |||
AK178223.1 | - | 4 | 308 | |||
AK180027.1 | - | 4 | 308 | |||
AK182665.1 | - | 4 | 306 | |||
AK183645.1 | - | 4 | 309 | |||
AK185811.1 | - | 3 | 284 | |||
AK187762.1 | - | 3 | 295 | |||
AK188385.1 | - | 4 | 306 | |||
AK207599.1 | - | 3 | 306 | |||
AK210479.1 | - | 3 | 275 | |||
AK220124.1 | - | 4 | 306 | |||
BC043072.1 | - | 6 | 806 | 6404165 |