Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dnaja2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019794.4 | NP_062768.1 | 3 | 374 | ||
GenBank mRNA | AB028853.1 | - | 3 | 375 | ||
AK031858.1 | - | 3 | 441 | |||
AK077672.1 | - | 3 | 445 | |||
AK083208.1 | - | 3 | 442 | |||
AK163255.1 | - | 3 | 373 | |||
AK169169.1 | - | 3 | 434 | |||
AK212721.1 | - | 1 | 85 | |||
BC003420.1 | - | 3 | 447 | 2582183 |