Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Noa1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019836.3 | NP_062810.1 | 3 | 1394 | ||
GenBank mRNA | AB041537.1 | - | 3 | 1366 | ||
AK011533.1 | - | 3 | 1386 | |||
AK017793.1 | - | 3 | 1367 | |||
AK019975.1 | - | 3 | 1376 | |||
AK128920.1 | - | 3 | 1366 | |||
AK139955.1 | - | 3 | 1338 | |||
AK157246.1 | - | 3 | 1394 | |||
AK163935.1 | - | 3 | 1368 | |||
AK183029.1 | - | 3 | 332 | |||
AK185846.1 | - | 3 | 239 | |||
BC005689.1 | - | 3 | 447 | |||
BC017154.1 | - | 3 | 1366 | 4910547 |