Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tdo2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019911.2 | NP_064295.2 | 5 | 360 | ||
GenBank mRNA | AB476989.1 | - | 5 | 834 | ||
AB476990.1 | - | 5 | 720 | |||
AK132271.1 | - | 5 | 383 | |||
BC018390.1 | - | 5 | 360 | 4159877 | ||
S56366.1 | - | 1 | 18 | |||
S56367.1 | - | 1 | 29 | |||
U24493.1 | - | 5 | 347 |