Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pex3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164195.1 | NP_001157667.1 | 9 | 1025 | ||
NM_019961.3 | NP_064345.1 | 9 | 1025 | |||
GenBank mRNA | AF152996.1 | - | 9 | 948 | ||
AK005315.1 | - | 9 | 881 | |||
AK005978.1 | - | 8 | 722 | |||
AK012827.1 | - | 9 | 1003 | |||
AK075820.1 | - | 9 | 940 | |||
AK083778.1 | - | 4 | 512 | |||
AK146934.1 | - | 9 | 1004 | |||
BC033415.1 | - | 9 | 907 | 5008068 | ||
BC037606.1 | - | 9 | 988 | 5361347 |