Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pdgfc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019971.2 | NP_064355.1 | 2, 3 | 1332 | ||
GenBank mRNA | AF117608.1 | - | 2, 3 | 508 | ||
AF266467.1 | - | 2, 3 | 1332 | |||
AF286725.1 | - | 2, 3 | 311 | |||
AK033734.1 | - | 2, 3 | 1061 | |||
AK042767.1 | - | 2, 3 | 1079 | |||
AK052947.1 | - | 2, 3 | 584 | |||
AK081347.1 | - | 3, 4 | 384 |