Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ick | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163780.1 | NP_001157252.1 | 2 | 542 | ||
NM_019987.2 | NP_064371.2 | 2 | 470 | |||
GenBank mRNA | AF225918.1 | - | 2 | 310 | ||
AK045950.1 | - | 2 | 469 | |||
AK087484.1 | - | 2 | 393 | |||
AK133537.1 | - | 2 | 409 | |||
AK154185.1 | - | 2 | 543 | |||
AK206365.1 | - | 1 | 123 | |||
BC028863.1 | - | 1 | 227 | 4224269 |