Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IFRD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006764.4 | NP_006755.4 | 3 | 539 | ||
GenBank mRNA | AK074763.1 | - | 3 | 356 | ||
BC001327.1 | - | 3 | 366 | 3456988 | ||
BC001676.2 | - | 3 | 351 | 2963821 | ||
BC007265.1 | - | 3 | 374 | 3138466 | ||
BC007437.2 | - | 3 | 399 | 2961581 | ||
BI825477.1 | - | 3 | 539 | |||
BT019999.1 | - | 3 | 218 | |||
U09585.1 | - | 3 | 216 | |||
Y12395.1 | - | 6 | 716 |