Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dhodh | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020046.3 | NP_064430.1 | 8, 9 | 1431 | ||
GenBank mRNA | AF029667.2 | - | 8, 9 | 1136 | ||
AK030798.1 | - | 1015 | ||||
BC019542.1 | - | 8, 9 | 1141 | 4039461 | ||
BC027829.1 | - | 8, 9 | 1392 | 5253723 | ||
BC045206.1 | - | 8, 9 | 1381 | 5253764 |