Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmem9b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020050.1 | NP_064434.1 | 3 | 334 | ||
GenBank mRNA | AK007430.1 | - | 3 | 302 | ||
AK009147.1 | - | 3 | 269 | |||
AK049882.1 | - | 3 | 421 | |||
AK077253.1 | - | 3 | 290 | |||
AK147127.1 | - | 3 | 350 | |||
AK201035.1 | - | 2 | 148 | |||
AK211319.1 | - | 2 | 148 | |||
BC002208.1 | - | 3 | 318 | 3490055 |