Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmem9b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020050.1 | NP_064434.1 | 3 | 346 | ||
GenBank mRNA | AK007430.1 | - | 3 | 314 | ||
AK009147.1 | - | 3 | 281 | |||
AK049882.1 | - | 3 | 433 | |||
AK077253.1 | - | 3 | 302 | |||
AK147127.1 | - | 3 | 362 | |||
AK201035.1 | - | 2 | 160 | |||
AK211319.1 | - | 2 | 160 | |||
BC002208.1 | - | 3 | 330 | 3490055 |