Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rufy3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001289774.1 | NP_001276703.1 | 5 | 883 | ||
NM_001289775.1 | NP_001276704.1 | 5 | 883 | |||
NM_001289776.1 | NP_001276705.1 | 6 | 1305 | |||
NM_001289777.1 | NP_001276706.1 | 7 | 1130 | |||
NM_027530.3 | NP_081806.1 | 5 | 1251 | |||
GenBank mRNA | AK013185.1 | - | 7 | 1131 | ||
AK018184.1 | - | 5 | 1229 | |||
AK083242.1 | - | 5 | 1186 | |||
AK148795.1 | - | 5 | 547 | |||
AK154941.1 | - | 5 | 884 | |||
AK160820.1 | - | 5 | 884 | |||
BC017648.1 | - | 5 | 640 | 4527858 | ||
BC049127.1 | - | 5 | 1190 | 6821567 | ||
BC058259.1 | - | 6 | 1024 | 6469137 | ||
CT010404.1 | - | 5 | 624 |