Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rgs7bp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029879.2 | NP_084155.2 | 6 | 1464 | ||
GenBank mRNA | AK028323.1 | - | 6 | 842 | ||
AK032138.1 | - | 6 | 1464 | |||
AK036240.1 | - | 6 | 1372 | |||
AK043529.1 | - | 3 | 268 | |||
AK046733.1 | - | 6 | 1093 | |||
AK137417.1 | - | 6 | 1338 | |||
BC118957.2 | - | 6 | 996 | 40093423 | ||
DQ104214.1 | - | 6 | 754 |